Tisk…

Nε-(tert-butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine 98%

Dodavatel: Thermo Scientific

Synonyms: Nepsilon-Boc-Nalpha-Fmoc-D-lysine, Fmoc-D-Lys(Boc)-OH

H28301.03 H28301.06
H28301.03EA 2040 CZK
H28301.03 H28301.06
Nε-(tert-butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine 98%
Nepsilon-(tert-butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine

Purity: 98% CAS No.: 92122-45-7 Molecular Formula: C26H32N2O6 Formula Weight: 468.54 Storage Temperature: 2°C to 8°C Melting Point: 135-139° Optical Rotation: 11° (c=1 in DMF) MDL No.: MFCD00065660

Vzorec: C₂₆H₃₂N₂O₆
MW: 468,55 g/mol
Bod tání: 135…139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065660
CAS číslo: 92122-45-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR