Tisk…

1-Benzyloxycarbonylpyrrolidine-3-carboxaldehyde 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: Benzyl 3-formylpyrrolidine-1-carboxylate

H28117.03 H28117.06
H28117.03EA 3050 CZK
H28117.03 H28117.06
1-Benzyloxycarbonylpyrrolidine-3-carboxaldehyde 97%
1-Benzyloxycarbonylpyrrolidine-3-carboxaldehyde

Purity: 97% CAS No.: 276872-86-7 Molecular Formula: C13H15NO3 Formula Weight: 233.27 Storage Temperature: 2°C to 8°C MDL No.: MFCD05861538

Vzorec: C₁₃H₁₅NO₃
MW: 233,27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05861538
CAS číslo: 276872-86-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR