4-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester 97%
Dodavatel: Thermo Scientific
Synonyms:
N-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
H27945.03
H27945.06
H27945.03EA
1690
CZK
H27945.03
H27945.06
4-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester 97%
4-(2-Methoxyethylaminocarbonyl)benzeneboronic acid pinacol ester
Purity: 97%
CAS No.: 1073353-60-2
Molecular Formula: C16H24BNO4
Formula Weight: 305.18
Melting Point: 73-76°
MDL No.: MFCD09266188
Vzorec:
C₁₆H₂₄BNO₄ MW: 305,18 g/mol Bod tání: 73…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD09266188 CAS číslo: 1073353-60-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...