2,3,4,5-Tetrahydro-1H-benzo[e][1,4]diazepine 95%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H27843.MD
H27843.03
H27843.MDEA
1230
CZK
H27843.MD
H27843.03
2,3,4,5-Tetrahydro-1H-benzo[e][1,4]diazepine 95%
2,3,4,5-Tetrahydro-1H-benzo[e][1,4]diazepine
Purity: 95%
CAS No.: 5946-39-4
Molecular Formula: C9H12N2
Formula Weight: 148.21
Melting Point: 96-100°
UN No.: UN3259
MDL No.: MFCD03789577
Vzorec:
C₉H₁₂N₂ MW: 148,21 g/mol Bod tání: 96…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD03789577 CAS číslo: 5946-39-4 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...