Tisk…

7-(Trifluoromethyl)quinoline 97%

Dodavatel: Thermo Scientific
Varování

H27771.MD H27771.03
H27771.MDEA 2040 CZK
H27771.MD H27771.03
7-(Trifluoromethyl)quinoline 97%
7-(Trifluoromethyl)quinoline

Purity: 97% CAS No.: 325-14-4 Molecular Formula: C10H6F3N Formula Weight: 197.16 Melting Point: 65-66° MDL No.: MFCD00833760

Vzorec: C₁₀H₆F₃N
MW: 197,16 g/mol
Bod tání: 65…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00833760
CAS číslo: 325-14-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR