α-(N,N-Dimethylamino)phenylacetonitrile 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H27631.14
H27631.06
H27631.14EA
3610
CZK
H27631.14
H27631.06
α-(N,N-Dimethylamino)phenylacetonitrile 97%
alpha-(N,N-Dimethylamino)phenylacetonitrile
Purity: 97%
CAS No.: 827-36-1
Molecular Formula: C10H12N2
Formula Weight: 160.22
Flash Point: 104°(219°F)
Refractive Index: 1.514
Boiling Point: 88-90°/2mm
UN No.: UN3276
MDL No.: MFCD00013810
Vzorec:
C₁₀H₁₂N₂ Bod varu: 88…90 °C (2 mmHg) Hustota: 0,997 Bod vzplanutí: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013810 CAS číslo: 827-36-1 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...