Tisk…

1-(1-Methylpiperidin-4-yl)piperazine 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 1-(1-Methyl-4-piperidinyl)piperazine

H27624.06 H27624.03
H27624.06EA 5630 CZK
H27624.06 H27624.03
1-(1-Methylpiperidin-4-yl)piperazine 98%
1-(1-Methylpiperidin-4-yl)piperazine

Purity: 98% CAS No.: 23995-88-2 Molecular Formula: C10H21N3 Formula Weight: 183.30 Refractive Index: 1.508 Melting Point: 29-32° UN No.: UN3259 MDL No.: MFCD01075186

Vzorec: C₁₀H₂₁N₃
Bod tání: 29…32 °C
Storage Temperature: Ambient
MDL Number: MFCD01075186
CAS číslo: 23995-88-2
UN: 3259
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR