Tisk…

4-N-Boc-amino-4-carboxytetrahydropyran 95%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4-((tert-Butoxycarbonyl)amino)tetrahydro-2H-pyran-4-carboxylic acid

H27578.03 H27578.MD
H27578.03EA 6590 CZK
H27578.03 H27578.MD
4-N-Boc-amino-4-carboxytetrahydropyran 95%
4-N-Boc-amino-4-carboxytetrahydropyran

Purity: 95% CAS No.: 172843-97-9 Molecular Formula: C11H19NO5 Formula Weight: 245.27 Melting Point: 157° dec. MDL No.: MFCD02683136

Vzorec: C₁₁H₁₉NO₅
Storage Temperature: Ambient
MDL Number: MFCD02683136
CAS číslo: 172843-97-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR