(R,R)-(-)-2,5-Bis(methoxymethyl)pyrrolidine 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H27577.ME
H27577.MC
H27577.MEEA
8170
CZK
H27577.ME
H27577.MC
(R,R)-(-)-2,5-Bis(methoxymethyl)pyrrolidine 97%
(R,R)-(-)-2,5-Bis(methoxymethyl)pyrrolidine
Purity: 97%
CAS No.: 90290-05-4
Molecular Formula: C8H17NO2
Formula Weight: 159.23
Refractive Index: 1.4490
Boiling Point: 106-108°/18mm
Optical Rotation: -8° (c=3 in ethanol)
MDL No.: MFCD00210015
Vzorec:
C₈H₁₇NO₂ MW: 159.23 g/mol Bod varu: 106…108 °C (18 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00210015 CAS číslo: 90290-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...