Tisk…

Ethyl-trans-4-bromocinnamate 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: (E)-Ethyl 3-(4-bromophenyl)acrylate

H27544.14 H27544.06
H27544.14EA 6150 CZK
H27544.14 H27544.06
Ethyl-trans-4-bromocinnamate 98%
Ethyl-trans-4-bromocinnamate

Purity: 98% CAS No.: 24393-53-1 Molecular Formula: C11H11BrO2 Formula Weight: 255.11 Flash Point: 110°(230°F) Refractive Index: 1.5970 Boiling Point: 180°/18mm MDL No.: MFCD00012226

Vzorec: C₁₁H₁₁BrO₂
MW: 255,11 g/mol
Bod varu: 180 °C (18 mmHg)
Hustota: 1,359
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012226
CAS číslo: 24393-53-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR