N,S-Dimethylisothiouronium hydriodide 98%
Dodavatel: Thermo Scientific
|
Varování
|
H27439.14
H27439.06
H27439.14EA
4120
CZK
H27439.14
H27439.06
N,S-Dimethylisothiouronium hydriodide 98%
N,S-Dimethylisothiouronium hydriodide
Purity: 98%
CAS No.: 41306-45-0
Molecular Formula: C3H8N2S.HI
Formula Weight: 232.07
Melting Point: 134-136°
MDL No.: MFCD00192092
Vzorec:
C₃H₈N₂S·HI MW: 232,07 g/mol Bod tání: 134…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00192092 CAS číslo: 41306-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...