4-Benzyloxy-6-chloropyrimidine 95%
Dodavatel: Thermo Scientific
|
Varování
|
H27390.MD
H27390.03
H27390.MDEA
4910
CZK
H27390.MD
H27390.03
4-Benzyloxy-6-chloropyrimidine 95%
4-Benzyloxy-6-chloropyrimidine
Purity: 95%
CAS No.: 405930-65-6
Molecular Formula: C11H9ClN2O
Formula Weight: 220.66
MDL No.: MFCD06798234
Vzorec:
C₁₁H₉ClN₂O MW: 220,66 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06798234 CAS číslo: 405930-65-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...