(R)-1-(Boc-amino)-2-propanol 98%
Dodavatel: Thermo Fisher Scientific
H27296.03
H27296.06
H27296.03EA
3810
CZK
H27296.03
H27296.06
(R)-1-(Boc-amino)-2-propanol 98%
(R)-1-(Boc-amino)-2-propanol
Purity: 98%
CAS No.: 119768-44-4
Molecular Formula: C8H17NO3
Formula Weight: 175.23
Refractive Index: 1.4460
Optical Rotation: -24° (c=1 in chloroform)
MDL No.: MFCD04974338
Vzorec:
C₈H₁₇NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD04974338 CAS číslo: 119768-44-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...