5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H27287.06
H27287.03
H27287.06EA
3850
CZK
H27287.06
H27287.03
5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile 97%
5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile
Purity: 97%
CAS No.: 51516-68-8
Molecular Formula: C10H7ClN4
Formula Weight: 218.65
Melting Point: 183-187°
MDL No.: MFCD00128298
Vzorec:
C₁₀H₇ClN₄ MW: 218,65 g/mol Bod tání: 183…187 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128298 CAS číslo: 51516-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...