Ethyl (E)-2-cyano-3-ethoxycrotonate 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H27244.14
H27244.06
H27244.14EA
3330
CZK
H27244.14
H27244.06
Ethyl (E)-2-cyano-3-ethoxycrotonate 98%
Ethyl (E)-2-cyano-3-ethoxycrotonate
Purity: 98%
CAS No.: 932750-29-3
Molecular Formula: C9H13NO3
Formula Weight: 183.21
Melting Point: 71-73°
UN No.: UN3439
MDL No.: MFCD01743485
Vzorec:
C₉H₁₃NO₃ MW: 183,21 g/mol Bod tání: 71…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD01743485 CAS číslo: 932750-29-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...