Tisk…

N-Methyl-tert-butylamine 97%

Dodavatel: Thermo Scientific
Nebezpečí

H27242.03 H27242.06
H27242.03EA 1120 CZK
H27242.03 H27242.06
N-Methyl-tert-butylamine 97%
N-Methyl-tert-butylamine

Purity: 97% CAS No.: 14610-37-8 Molecular Formula: C5H13N Formula Weight: 87.16 Flash Point: -18°(0°F) Refractive Index: 1.3950 Odour: Amine-like Boiling Point: 67-69° UN No.: UN2733 MDL No.: MFCD00042853

Vzorec: C₅H₁₃N
MW: 87,166 g/mol
Bod varu: 67…69 °C
Hustota: 0,727
Bod vzplanutí: –18 °C (0 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042853
CAS číslo: 14610-37-8
EINECS: 238-646-2
UN: 2733
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR