Tisk…

Methyl-7-bromoheptanoate 98%

Dodavatel: Thermo Scientific

Synonyms: Methyl 7-bromoheptanoate

H27234.14 H27234.06
H27234.14EA 8100 CZK
H27234.14 H27234.06
Methyl-7-bromoheptanoate 98%
Methyl-7-bromoheptanoate

Purity: 98% CAS No.: 54049-24-0 Molecular Formula: Br(CH2)6CO2CH3 Formula Weight: 223.11 MDL No.: MFCD02258672

Vzorec: C₈H₁₅BrO₂
MW: 223,12 g/mol
Hustota: 1,257
Storage Temperature: Ambient
MDL Number: MFCD02258672
CAS číslo: 54049-24-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR