6-Acetyl-2(3H)-benzothiazolone 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H27154.09
H27154.03
H27154.09EA
4120
CZK
H27154.09
H27154.03
6-Acetyl-2(3H)-benzothiazolone 97%
6-Acetyl-2(3H)-benzothiazolone
Purity: 97%
CAS No.: 133044-44-7
Molecular Formula: C9H7NO2S
Formula Weight: 193.22
Melting Point: 190-194°
MDL No.: MFCD02660572
Vzorec:
C₉H₇NO₂S Bod tání: 190…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD02660572 CAS číslo: 133044-44-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...