2-Chloro-6-methoxy-3-quinoline carboxaldehyde 99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Chloro-6-methoxyquinoline-3-carbaldehyde
H26973.14
H26973.06
H26973.14EA
7970
CZK
H26973.14
H26973.06
2-Chloro-6-methoxy-3-quinoline carboxaldehyde 99%
2-Chloro-6-methoxy-3-quinoline carboxaldehyde
Purity: 99%
CAS No.: 73568-29-3
Molecular Formula: C11H8ClNO2
Formula Weight: 221.64
Flash Point: -9°(16°F)
Melting Point: 146-151°
MDL No.: MFCD00160585
Vzorec:
C₁₁H₈ClNO₂ MW: 221,64 g/mol Bod tání: 146…151 °C Bod vzplanutí: –9 °C (16 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00160585 CAS číslo: 73568-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...