Tisk…

2'-Iodo-1,1':3',1''-terphenyl ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2'-Iodo-1,1':3',1''-terphenyl, 1-Iodo-2,6-diphenylbenzene, 2'-Iodo-m-terphenyl

H26704.03 H26704.06
H26704.03EA 3050 CZK
H26704.03 H26704.06
2'-Iodo-1,1':3',1''-terphenyl ≥99%
2'-Iodo-1,1':3',1''-terphenyl

Purity: 99% CAS No.: 82777-09-1 Molecular Formula: C18H13I Formula Weight: 356.21 Melting Point: 113-116° MDL No.: MFCD00185026

Vzorec: C₁₈H₁₃I
MW: 356,21 g/mol
Bod tání: 113…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00185026
CAS číslo: 82777-09-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR