2-Valerylfuran 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H26491.09
H26491.18
H26491.09EA
1850
CZK
H26491.09
H26491.18
2-Valerylfuran 97%
2-Valerylfuran
Purity: 97%
CAS No.: 3194-17-0
Molecular Formula: C9H12O2
Formula Weight: 152.20
Refractive Index: 1.4920
MDL No.: MFCD00053147
Vzorec:
C₉H₁₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00053147 CAS číslo: 3194-17-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...