Tisk…

Hypericin 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1,3,4,6,8,13-Hexahydroxy-10,11-dimethylphenanthro[1,10,9,8-opqra]perylene-7,14-dione P-conformer

H26425.MB H26425.MC
H26425.MBEA 14010 CZK
H26425.MB H26425.MC
Hypericin 98%
Hypericin

Purity: 98% CAS No.: 548-04-9 Molecular Formula: C30H16O8 Formula Weight: 504.44 Storage Temperature: -30°C to -10°C Melting Point: ca 320° dec. Beilstein Reference: 1917913 MDL No.: MFCD00016683

Vzorec: C₃₀H₁₆O₈
MW: 504,45 g/mol
Bod tání: ∼ 320 °C (rozkl.)
Storage Temperature: Freezer
MDL Number: MFCD00016683
CAS číslo: 548-04-9
EINECS: 208-941-0
Merck Index: 14,4863

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR