Tisk…

Dimethyl (4-fluorobenzyl)malonate 97%

Dodavatel: Thermo Fisher Scientific

H26310.03 H26310.06
H26310.03EA 1410 CZK
H26310.03 H26310.06
Dimethyl (4-fluorobenzyl)malonate 97%
Dimethyl (4-fluorobenzyl)malonate

Purity: 97% CAS No.: 252938-22-0 Molecular Formula: C12H13FO4 Formula Weight: 240.23 MDL No.: MFCD01934558

Vzorec: C₁₂H₁₃FO₄
Storage Temperature: Ambient
MDL Number: MFCD01934558
CAS číslo: 252938-22-0
EINECS: None

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR