4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid 97%
Dodavatel: Thermo Scientific
|
Varování
|
H26154.MD
H26154.03
H26154.MDEA
2700
CZK
H26154.MD
H26154.03
4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid 97%
4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid
Purity: 97%
CAS No.: 878572-17-9
Molecular Formula: C8H3F4NO4
Formula Weight: 253.11
Melting Point: 120-124°
MDL No.: MFCD08282778
Vzorec:
C₈H₃F₄NO₄ MW: 253,11 g/mol Bod tání: 120…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD08282778 CAS číslo: 878572-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...