Acetylacetonato(1,5-cyclooctadiene)rhodium(I)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(Acetylacetonato)(1,5-cyclooctadiene)rhodium(I)
H26083.03
H26083.MD
H26083.03EA
30000
CZK
H26083.03
H26083.MD
Acetylacetonato(1,5-cyclooctadiene)rhodium(I)
Acetylacetonato(1,5-cyclooctadiene)rhodium(I)
CAS No.: 12245-39-5
Molecular Formula: C13H19O2Rh
Formula Weight: 310.19
Melting Point: 136-140°
MDL No.: MFCD00075046
Vzorec:
C₁₃H₂₀O₂Rh MW: 311.207 g/mol Bod tání: 136…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075046 CAS číslo: 12245-39-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...