Tisk…

3,6-Dimethyl-4-hydroxy-2-pyrone 97%

Dodavatel: Thermo Scientific
Varování

H26074.06 H26074.03
H26074.06EA 20420 CZK
H26074.06 H26074.03
3,6-Dimethyl-4-hydroxy-2-pyrone 97%
3,6-Dimethyl-4-hydroxy-2-pyrone

Purity: 97% CAS No.: 5192-62-1 Molecular Formula: C7H8O3 Formula Weight: 140.14 Melting Point: 209-211° MDL No.: MFCD00090424

Vzorec: C₇H₈O₃
MW: 140,14 g/mol
Bod tání: 209…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00090424
CAS číslo: 5192-62-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR