Tisk…

3,3,3-Trifluoro-2,2-dimethylpropionic acid 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 3,3,3-Trifluoro-2,2-dimethylpropanoic acid

H26053.MD H26053.03
H26053.MDEA 2150 CZK
H26053.MD H26053.03
3,3,3-Trifluoro-2,2-dimethylpropionic acid 97%
3,3,3-Trifluoro-2,2-dimethylpropionic acid

Purity: 97% CAS No.: 889940-13-0 Molecular Formula: C5H7F3O2 Formula Weight: 156.12 Melting Point: 66-73° Boiling Point: 76-77°/14mm UN No.: UN3261 MDL No.: MFCD08445819

Vzorec: C₅H₇F₃O₂
MW: 156,11 g/mol
Bod varu: 76…77 °C (14 mmHg)
Bod tání: 66…73 °C
Storage Temperature: Ambient
MDL Number: MFCD08445819
CAS číslo: 889940-13-0
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR