Tisk…

4-(4-Fluorophenyl)butyric acid 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-(4-Fluorophenyl)butyric Acid

H26035.MD H26035.03
H26035.MDEA 855 CZK
H26035.MD H26035.03
4-(4-Fluorophenyl)butyric acid 97%
4-(4-Fluorophenyl)butyric acid

Purity: 97% CAS No.: 589-06-0 Molecular Formula: C10H11FO2 Formula Weight: 182.20 Melting Point: 44-46° MDL No.: MFCD03788503

Vzorec: C₁₀H₁₁FO₂
MW: 182,2 g/mol
Bod tání: 44…46 °C
Storage Temperature: Ambient
MDL Number: MFCD03788503
CAS číslo: 589-06-0
EINECS: 209-631-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR