5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin 95%
Dodavatel: Thermo Fisher Scientific
H25946.09
H25946.03
H25946.09EA
10510
CZK
H25946.09
H25946.03
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin 95%
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin
Purity: 95%
CAS No.: 22112-78-3
Molecular Formula: C48H38N4O4
Formula Weight: 734.87
Storage Temperature: 2°C to 8°C
MDL No.: MFCD09909566
Vzorec:
C₄₈H₃₈N₄O₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD00012072 CAS číslo: 22112-78-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...