Tisk…

Methyl-2-oxo-2H-pyran-3-carboxylate 98%

Dodavatel: Thermo Fisher Scientific

H25932.06 H25932.03
H25932.06EA 3810 CZK
H25932.06 H25932.03
Methyl-2-oxo-2H-pyran-3-carboxylate 98%
Methyl-2-oxo-2H-pyran-3-carboxylate

Purity: 98% CAS No.: 25991-27-9 Molecular Formula: C7H6O4 Formula Weight: 154.12 Melting Point: 74-77° Boiling Point: 146-148°/0.75mm Beilstein Reference: 1424749 MDL No.: MFCD00075443

Vzorec: C₇H₆O₄
Bod varu: 146…148 °C (0,75 mmHg)
Bod tání: 74…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00075443
CAS číslo: 25991-27-9
EINECS: 247-397-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR