3-Methylthiopropylamine 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H25846.06
H25846.03
H25846.06EA
5500
CZK
H25846.06
H25846.03
3-Methylthiopropylamine 97%
3-Methylthiopropylamine
Purity: 97%
CAS No.: 4104-45-4
Molecular Formula: C4H11NS
Formula Weight: 105.20
Flash Point: 62°(144°F)
Refractive Index: 1.4940
Odour: Stench
Boiling Point: 169°
UN No.: UN2735
MDL No.: MFCD00041898
Vzorec:
C₄H₁₁NS Bod varu: 169 °C Hustota: 0,938 Bod vzplanutí: 62 °C (144 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041898 CAS číslo: 4104-45-4 EINECS: 223-875-2 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...