N-Methyl-N-methoxydifluoroacetamide 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2,2-Difluoro-N-methoxy-N-methylacetamide
H25833.06
H25833.03
H25833.06EA
9650
CZK
H25833.06
H25833.03
N-Methyl-N-methoxydifluoroacetamide 95%
N-Methyl-N-methoxydifluoroacetamide
Purity: 95%
CAS No.: 142492-01-1
Molecular Formula: C4H7F2NO2
Formula Weight: 139.10
Refractive Index: 1.4020
Boiling Point: 40-41°/0.7mm
MDL No.: MFCD06248152
Vzorec:
C₄H₇F₂NO₂ MW: 139,1 g/mol Bod varu: 40…41 °C (0,7 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD06248152 CAS číslo: 142492-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...