Ethyl-5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate ≥98%
Dodavatel: Thermo Scientific
H25763.03
H25763.MD
H25763.03EA
5220
CZK
H25763.03
H25763.MD
Ethyl-5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate ≥98%
Ethyl-5-phenoxycarbonylamino-1,2,3-thiadiazole-4-carboxylate
Purity: 98+%
CAS No.: 2037-81-2
Molecular Formula: C12H11N3O4S
Formula Weight: 293.30
Melting Point: 153-155°
MDL No.: MFCD00100230
Vzorec:
C₁₂H₁₁N₃O₄S MW: 293,30 g/mol Bod tání: 153…155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00100230 CAS číslo: 2037-81-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...