Tisk…

Ethyl-2,4-dioxo-5-methylhexanoate 95%

Dodavatel: Thermo Scientific

Synonyms: Ethyl 5-methyl-2,4-dioxohexanoate

B25735.06 B25735.14
B25735.06EA 1670 CZK
B25735.06 B25735.14
Ethyl-2,4-dioxo-5-methylhexanoate 95%
Ethyl-2,4-dioxo-5-methylhexanoate

Purity: 95% CAS No.: 64195-85-3 Molecular Formula: C9H14O4 Formula Weight: 186.21 MDL No.: MFCD07323638

Vzorec: C₉H₁₄O₄
MW: 186,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07323638
CAS číslo: 64195-85-3
EINECS: 264-728-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR