Dimethyl iminodiacetate hydrochloride 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B25729.06
B25729.14
B25729.06EA
2710
CZK
B25729.06
B25729.14
Dimethyl iminodiacetate hydrochloride 98%
Dimethyl iminodiacetate hydrochloride
Purity: 98%
CAS No.: 39987-25-2
Molecular Formula: C6H11NO4.HCl
Formula Weight: 197.62
Melting Point: ca 170°
Beilstein Reference: 3703236
MDL No.: MFCD00216724
Vzorec:
C₆H₁₁NO₄·HCl Bod tání: ∼ 170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216724 CAS číslo: 39987-25-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...