Tisk…

Ethyl-2-((diphenylmethylene)amino)acetate 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: Ethyl (diphenylmethylenamino)acetate, Ethyl 2-((diphenylmethylene)amino)acetate, Ethyl-2-((diphenylmethylene)amino)acetate, Ethyl N-(diphenylmethylene)glycinate

B25580.06 B25580.14
B25580.06EA 2560 CZK
B25580.06 B25580.14
Ethyl-2-((diphenylmethylene)amino)acetate 98%
N-(Diphenylmethylene)glycine ethyl ester

Purity: 98% CAS No.: 69555-14-2 Molecular Formula: C17H17NO2 Formula Weight: 267.33 Flash Point: 110°(230°F) Melting Point: 51-55° MDL No.: MFCD00010590

Vzorec: C₁₇H₁₇NO₂
Bod tání: 51…55 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010590
CAS číslo: 69555-14-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR