Methyl (4-fluorobenzoyl)acetate 95%
Dodavatel: Thermo Fisher Scientific
Synonyms:
3-(4-Fluorophenyl)-3-oxopropionic Acid Methyl Ester, Methyl 4-fluorobenzoylacetate, (4-Fluorobenzoyl)acetic Acid Methyl Ester, Methyl 3-(4-Fluorophenyl)-3-oxopropionate
B25546.03
B25546.09
B25546.03EA
1260
CZK
B25546.03
B25546.09
Methyl (4-fluorobenzoyl)acetate 95%
Methyl (4-fluorobenzoyl)acetate
Purity: 95%
CAS No.: 63131-29-3
Molecular Formula: C10H9FO3
Formula Weight: 196.18
Flash Point:
Vzorec:
C₁₀H₉FO₃ MW: 196.18 g/mol Bod varu: 91 °C (5 mmHg) Hustota: 1.228 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000355 CAS číslo: 63131-29-3 EINECS: 263-889-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...