4'-(4-Methoxyphenyl)-2,2':6',2''-terpyridine 98%
Dodavatel: Thermo Scientific
|
Varování
|
B25486.02
B25486.77
B25486.02EA
2180
CZK
B25486.02
B25486.77
4'-(4-Methoxyphenyl)-2,2':6',2''-terpyridine 98%
4'-(4-Methoxyphenyl)-2,2':6',2''-terpyridine
Purity: 98%
CAS No.: 13104-56-8
Molecular Formula: C22H17N3O
Formula Weight: 339.40
Melting Point: 161-163°
MDL No.: MFCD06796987
Vzorec:
C₂₂H₁₇N₃O MW: 339,40 g/mol Bod tání: 161…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD06796987 CAS číslo: 13104-56-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...