2-Methylpropane-2-sulphinamide 97%
Dodavatel: Thermo Scientific
B25385.03
B25385.06
B25385.03EA
1510
CZK
B25385.03
B25385.06
2-Methylpropane-2-sulphinamide 97%
2-Methylpropane-2-sulphinamide
Purity: 97%
CAS No.: 146374-27-8
Molecular Formula: (CH3)3CS(O)NH2
Formula Weight: 121.20
Melting Point: 97-101°
MDL No.: MFCD05861480
Vzorec:
C₄H₁₁NOS MW: 121,20 g/mol Bod tání: 97…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD01863616 CAS číslo: 146374-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...