Tisk…

Quinoline-N-oxide hydrate, (∼20% H₂O) 97% (dry weight)

Dodavatel: Thermo Scientific

Synonyms: Quinoline 1-oxide hydrate

B25209.06 B25209.14
B25209.06EA 1730 CZK
B25209.06 B25209.14
Quinoline-N-oxide hydrate, (∼20% H₂O) 97% (dry weight)
Quinoline-N-oxide hydrate

Purity: 97% CAS No.: 64201-64-5 Molecular Formula: C9H7NO.xH2O Formula Weight: 145.16(anhy) Flash Point: 110°(230°F) Melting Point: ca 50° Beilstein Reference: 5989589 MDL No.: MFCD00149472

Vzorec: C₉H₇NO·xH₂O
MW: 145,16 g/mol (anhy)
Bod tání: ∼ 50 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00149472
CAS číslo: 64201-64-5
EINECS: 216-560-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR