Tisk…

4-(4-Hydroxy-3-methoxyphenyl)-2-butanone 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: Zingerone, Vanillylacetone

B25150.03 B25150.06 B25150.14
B25150.03EA 1070 CZK
B25150.03 B25150.06 B25150.14
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone 97%
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone

Purity: 97% CAS No.: 122-48-5 Molecular Formula: C11H14O3 Formula Weight: 194.23 Melting Point: 39-42° MDL No.: MFCD00048232

Vzorec: C₁₁H₁₄O₃
MW: 194,23 g/mol
Bod tání: 39…42 °C
Storage Temperature: Ambient
MDL Number: MFCD00048232
CAS číslo: 122-48-5
EINECS: 204-548-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR