4-Pyridylacetonitrile hydrochloride 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2-(Pyridin-4-yl)acetonitrile hydrochloride
B25095.03
B25095.06
B25095.14
B25095.03EA
1850
CZK
B25095.03
B25095.06
B25095.14
4-Pyridylacetonitrile hydrochloride 98%
4-Pyridylacetonitrile hydrochloride
Purity: 98%
CAS No.: 92333-25-0
Molecular Formula: C7H6N2.HCl
Formula Weight: 154.60
Melting Point: ca 260° dec.
UN No.: UN3439
MDL No.: MFCD00012825
Vzorec:
C₇H₇ClN₂ MW: 154.6 g/mol Bod tání: ∼ 260 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00012825 CAS číslo: 92333-25-0 UN: 3439 ADR: 6.1 III |