Ethyl-2-cyclohexylidenacetate 98%
Dodavatel: Thermo Fisher Scientific
B25073.06
B25073.14
B25073.22
B25073.06EA
1850
CZK
B25073.06
B25073.14
B25073.22
Ethyl-2-cyclohexylidenacetate 98%
Ethyl-2-cyclohexylidenacetate
Purity: 98%
CAS No.: 1552-92-7
Molecular Formula: C10H16O2
Formula Weight: 168.24
Boiling Point: 48-49°/0.02mm
MDL No.: MFCD00017627
Vzorec:
C₁₀H₁₆O₂ Bod varu: 48…49 °C (0,02 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00963857 CAS číslo: 1552-92-7 EINECS: 216-297-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...