Tisk…

3-Thien-3-ylacrylic acid 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-(Thien-3-yl)prop-2-enoic acid, 3-(2-Carboxyethenyl)thiophene, (E)-3-(Thiophen-3-yl)acrylic acid, 3-(Thien-3-yl)acrylic acid

B25017.06 B25017.14
B25017.06EA 2710 CZK
B25017.06 B25017.14
3-Thien-3-ylacrylic acid 98%
3-Thien-3-ylacrylic acid

Purity: 98% CAS No.: 102696-71-9 Molecular Formula: C7H6O2S Formula Weight: 154.19 Melting Point: 150-154° Beilstein Reference: 112245 MDL No.: MFCD00014535

Vzorec: C₇H₆O₂S
MW: 154,19 g/mol
Bod tání: 150…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00014535
CAS číslo: 102696-71-9
EINECS: 214-800-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR