Tisk…

Anthraquinone-2-sulphonic acid sodium salt, (H₂O 4-6%) 97% (dry weight)

Dodavatel: Thermo Fisher Scientific
Varování

B24969.0E B24969.22 B24969.36
B24969.0EEA 9580 CZK
B24969.0E B24969.22 B24969.36
Anthraquinone-2-sulphonic acid sodium salt, (H₂O 4-6%) 97% (dry weight)
Anthraquinone-2-sulphonic acid sodium salt

Purity: 97% CAS No.: 131-08-8 Molecular Formula: C14H7NaO5S.xH2O Formula Weight: 310.26(anhy) Melting Point: 320° Beilstein Reference: 3580452 MDL No.: MFCD00149068

Vzorec: C₁₄H₇NaO₅S·xH₂O
Bod tání: >320 °C
Storage Temperature: Ambient
MDL Number: MFCD00149068
CAS číslo: 131-08-8
EINECS: 205-009-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR