4-Hydroxybenzoylacrylic acid 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B24519.06
B24519.14
B24519.06EA
5150
CZK
B24519.06
B24519.14
4-Hydroxybenzoylacrylic acid 97%
4-Hydroxybenzoylacrylic acid
Purity: 97%
CAS No.: 24849-48-7
Molecular Formula: C10H8O4
Formula Weight: 192.17
Melting Point: 197-198°
MDL No.: MFCD00460607
Vzorec:
C₁₀H₈O₄ Bod tání: 197…198 °C Storage Temperature: Ambient |
MDL Number:
MFCD00460607 CAS číslo: 24849-48-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...