Tisk…

Potassium sulphamate 98%

Dodavatel: Thermo Fisher Scientific
Varování

B24447.14 B24447.22 B24447.36
B24447.14EA 1500 CZK
B24447.14 B24447.22 B24447.36
Potassium sulphamate 98%
Potassium sulphamate

Purity: 98% CAS No.: 13823-50-2 Molecular Formula: H2KNO3S Formula Weight: 135.18 Physical Form: Crystalline solid Sensitivity: Hygroscopic Appearance: White MDL No.: MFCD00059941

Vzorec: H₃NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00059941
CAS číslo: 13823-50-2
EINECS: 237-515-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR