β-D-Ribofuranose-1,2,3,5-tetraacetate ≥98%
Dodavatel: Thermo Scientific
Synonyms:
Tetra-O-acetyl-β-D-ribofuranose, (2S,3R,4R,5R)-5-(Acetoxymethyl)tetrahydrofuran-2,3,4-triyl triacetate
B24426.06
B24426.14
B24426.22
B24426.06EA
1300
CZK
B24426.06
B24426.14
B24426.22
β-D-Ribofuranose-1,2,3,5-tetraacetate ≥98%
beta-D-Ribofuranose-1,2,3,5-tetraacetate
Purity: 98+%
CAS No.: 13035-61-5
Molecular Formula: C13H18O9
Formula Weight: 318.28
Melting Point: 80-83°
Beilstein Reference: 94078
Optical Rotation: -13+/-0.5° (c=2.4 in chloroform)
MDL No.: MFCD00005358
Vzorec:
C₁₃H₁₈O₉ MW: 318,28 g/mol Bod tání: 80…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005358 CAS číslo: 13035-61-5 EINECS: 235-898-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...