Tisk…

Cobalt(II) gluconate hydrate

Dodavatel: Thermo Scientific
Nebezpečí

B24293.14 B24293.22
B24293.14EA 1580 CZK
B24293.14 B24293.22
Cobalt(II) gluconate hydrate
Cobalt(II) gluconate hydrate

CAS No.: 71957-08-9 Molecular Formula: C12H22CoO14.xH2O Formula Weight: 449.23(anhy) Melting Point: 300° MDL No.: MFCD02684006

Vzorec: C₁₂H₂₂CoO₁₄·xH₂O
MW: 449,23 g/mol (anhy)
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD02684006
CAS číslo: 1809162-99-9
EINECS: 276-206-1
UN: 3288
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR