Zincon sodium salt
Dodavatel: Thermo Scientific
|
Varování
|
B24256.03
B24256.06
B24256.14
B24256.03EA
1180
CZK
B24256.03
B24256.06
B24256.14
Zincon sodium salt
Zincon sodium salt
CAS No.: 62625-22-3
Molecular Formula: C20H15N4NaO6S
Formula Weight: 462.42
Melting Point: ca 203° dec.
Beilstein Reference: 3872699
MDL No.: MFCD00064385
Vzorec:
C₂₀H₁₅N₄NaO₆S MW: 462,412 g/mol Bod tání: ∼ 203 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00064385 CAS číslo: 62625-22-3 EINECS: 263-651-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...